Anti-CD44 Mouse Monoclonal Antibody

Supplier: ProSci
51-648
10077-184EA 491.23 USD
10077-184
Anti-CD44 Mouse Monoclonal Antibody
CD44 antibody can be used in Western Blot, and immunohistochemistry starting at 10 ?g/mL.

10 mM PBS, pH 7.4, BSA, sodium azide.

Store CD44 antibody at 2 °C to 8 °C degrees. Do not freeze.

Type:
Antigen: CD44
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR