Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TRPM7 Mouse Monoclonal Antibody

Supplier: Prosci
51-634
10077-158
Anti-TRPM7 Mouse Monoclonal Antibody
TRPM7 antibody can be used in immunohistochemistry starting at 10 μg/mL, and flow cytometry starting at 0.5 µg/1E6 cells.

Protein G Column

PBS, pH 7.4, 50% glycerol, 0.09% sodium azide

Store TRPM7 antibody at -20 °C.

Type:
Antigen: TRPM7
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now


 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty