Anti-SERPINC1 Mouse Monoclonal Antibody

Supplier: ProSci
51-622
10077-140EA 674.53 USD
10077-140
Anti-SERPINC1 Mouse Monoclonal Antibody
Antibodies
Antithrombin-III antibody can be used in Western Blot, and immunohistochemistry starting at 1:50.

HEPES with 0.15M NaCl, 0.01% BSA, 0.03% sodium azide, and 50% glycerol

Store Antithrombin-III antibody at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: SERPINC1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR