Anti-COPS5 Mouse Monoclonal Antibody [clone: [8H8.5]]

Supplier: Prosci

51-620
10077-136EA 582.9 USD
10077-136
Anti-COPS5 Mouse Monoclonal Antibody [clone: [8H8.5]]
Antibodies
COPS5 antibody can be used in ELISA, and immunohistochemistry starting at 10 ?g/mL.

Phosphate buffer, pH 7.4. Carrier, preservative free.

COPS5 antibody should be stored at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: COPS5
Clonality: Monoclonal
Clone: [8H8.5]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR