Anti-TUBB2A Mouse Monoclonal Antibody [clone: [2B2]]

Supplier: ProSci
51-610
10077-118EA 749.47 USD
10077-118
Anti-TUBB2A Mouse Monoclonal Antibody [clone: [2B2]]
Antibodies
TUBB2A antibody can be used in Western Blot, and immunohistochemistry starting at 1:50.

PBS, pH 7.2

Store TUBB2A antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: TUBB2A
Clonality: Monoclonal
Clone: [2B2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR