Anti-RNF144B Mouse Monoclonal Antibody [clone: [4F1]]

Supplier: Prosci

51-606
10077-110EA 647.66 USD
10077-110
Anti-RNF144B Mouse Monoclonal Antibody [clone: [4F1]]
Antibodies
RNF144B antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

Store RNF144B antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: RNF144B
Clonality: Monoclonal
Clone: [4F1]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR