Anti-ADAMTS17 Mouse Monoclonal Antibody

Supplier: Prosci

51-604
10077-106EA 624.31 USD InStock
10077-106
Anti-ADAMTS17 Mouse Monoclonal Antibody
Antibodies
ADAMTS17 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 15 ?g/mL, immunocytochemistry, and immunoprecipitation.

PBS, pH 7.2

Store ADAMTS17 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: ADAMTS17
Clonality: Monoclonal
Clone: [3B7]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR