Anti-AKR1B10 Mouse Monoclonal Antibody [clone: [1A6]]

Supplier: ProSci
51-587
10079-560EA 749.47 USD
10079-560
Anti-AKR1B10 Mouse Monoclonal Antibody [clone: [1A6]]
Antibodies
AKR1B10 antibody can be used in Western Blot, immunohistochemistry starting at 10 ?g/mL, and immunocytochemistry.

PBS, pH 7.2

Store AKR1B10 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: AKR1B10
Clonality: Monoclonal
Clone: [1A6]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR