Anti-CCND1 Mouse Monoclonal Antibody [clone: [CD1.1]]

Supplier: ProSci
51-565
10079-516EA 674.53 USD
10079-516
Anti-CCND1 Mouse Monoclonal Antibody [clone: [CD1.1]]
Antibodies
Cyclin D1 antibody can be used in Western Blot, and immunohistochemistry starting at 5 ?g/mL.

PBS, 15 mM sodium azide, pH 7.4

Cyclin D1 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: CCND1
Clonality: Monoclonal
Clone: [CD1.1]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR