Anti-SMARCA4 Mouse Monoclonal Antibody [clone: [BRG-01]]

Supplier: ProSci
51-564
10079-514EA 698.16 USD
10079-514
Anti-SMARCA4 Mouse Monoclonal Antibody [clone: [BRG-01]]
Antibodies
BRG1 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 ?g/mL.

PBS, 15 mM sodium azide, pH 7.4

BRG1 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: SMARCA4
Clonality: Monoclonal
Clone: [BRG-01]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR