Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSP90AB1 Mouse Monoclonal Antibody

Supplier: Prosci
51-563
10079-512
Anti-HSP90AB1 Mouse Monoclonal Antibody
HSP90AB1 antibody can be used in immunohistochemistry, immunohistochemistry starting at 1:50, immunoprecipitation, and flow cytometry.

Protein A Column

15 mM sodium azide, 0.2% BSA, PBS

HSP90AB1 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type:
Antigen: HSP90AB1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Dog, Xenopus, Gorilla, Goat, Horse, Human, Monkey, Mouse, Orangutan, Pig, Rabbit, Rat

Order Now


 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty