Anti-NEFH Mouse Monoclonal Antibody [clone: [NF-05]]

Supplier: Prosci

51-560
10079-506EA 561.89 USD
10079-506
Anti-NEFH Mouse Monoclonal Antibody [clone: [NF-05]]
Antibodies
NF-H antibody can be used in Western Blot, immunohistochemistry starting at 5 ?g/mL, and immunoprecipitation.

PBS, 15 mM sodium azide, pH 7.4

Store NF-H antibody at 4 °C. Do not freeze.

Type: Primary
Antigen: NEFH
Clonality: Monoclonal
Clone: [NF-05]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR