Anti-KIF5B Mouse Monoclonal Antibody [clone: [KN-03]]

Supplier: ProSci
51-559
10079-504EA 698.16 USD
10079-504
Anti-KIF5B Mouse Monoclonal Antibody [clone: [KN-03]]
Antibodies
KImmunofluorescence5B antibody can be used in immunohistochemistry starting at 10 ?g/mL.

PBS, 15 mM sodium azide, approx., pH 7.4.

Store KIF5B antibody at 4 °C. Do not freeze.

Type: Primary
Antigen: KIF5B
Clonality: Monoclonal
Clone: [KN-03]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR