Anti-TUBB3 Mouse Monoclonal Antibody [clone: [TU-20]]

Supplier: Prosci

51-557
10077-084EA 582.9 USD
10077-084
Anti-TUBB3 Mouse Monoclonal Antibody [clone: [TU-20]]
Antibodies
Beta III Tubulin antibody can be used in ELISA, Western Blot starting at 1 ?g/mL, and immunohistochemistry starting at 10 - 20 ?g/mL.

PBS, 15 mM sodium azide, approx., pH 7.4.

Beta III Tubulin antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: TUBB3
Clonality: Monoclonal
Clone: [TU-20]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR