Anti-YAP1 Mouse Monoclonal Antibody [clone: [2F12]]

Supplier: ProSci
51-552
10077-076EA 760.5 USD
10077-076
Anti-YAP1 Mouse Monoclonal Antibody [clone: [2F12]]
Antibodies
YAP1 antibody can be used in ELISA starting at 1:62500, Western Blot starting at 2.5 ?g/mL, and immunohistochemistry starting at 20 ?g/mL.

PBS, pH 7.2

Store YAP1 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: YAP1
Clonality: Monoclonal
Clone: [2F12]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR