Anti-BANF1 Mouse Monoclonal Antibody [clone: [3F10-4G12]]

Supplier: ProSci
51-534
10077-040EA 749.47 USD
10077-040
Anti-BANF1 Mouse Monoclonal Antibody [clone: [3F10-4G12]]
Antibodies
BANF1 antibody can be used in Western Blot starting at 1: 500 - 1: 2000, and immunohistochemistry starting at 1:800.

PBS, pH 7.2

Store BANF1 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: BANF1
Clonality: Monoclonal
Clone: [3F10-4G12]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR