Anti-CDKL1 Mouse Monoclonal Antibody [clone: [2H6]]

Supplier: ProSci
51-533
10077-038EA 749.24 USD
10077-038
Anti-CDKL1 Mouse Monoclonal Antibody [clone: [2H6]]
Antibodies
CDKL1 antibody can be used in ELISA starting at 1: 10000, Western Blot starting at 1: 500 - 1: 1000, and immunohistochemistry starting at 1: 1000 - 1: 2000.

PBS, pH 7.2

Store CDKL1 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: CDKL1
Clonality: Monoclonal
Clone: [2H6]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR