Anti-TOP2A Mouse Monoclonal Antibody [clone: [1E2]]

Supplier: Prosci

51-523
10073-274EA 624.31 USD
10073-274
Anti-TOP2A Mouse Monoclonal Antibody [clone: [1E2]]
Antibodies
TOP2A antibody can be used in Western Blot, immunohistochemistry starting at 1:50, and immunofluorescence.

PBS, pH 7.2

Store TOP2A antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: TOP2A
Clonality: Monoclonal
Clone: [1E2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR