Anti-TLN1 Mouse Monoclonal Antibody [clone: [5C1]]

Supplier: ProSci
51-521
10073-270EA 760.5 USD
10073-270
Anti-TLN1 Mouse Monoclonal Antibody [clone: [5C1]]
Antibodies
Talin antibody can be used in Western Blot, immunohistochemistry starting at 1:100, and immunofluorescence.

PBS, pH 7.2

Store Talin antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: TLN1
Clonality: Monoclonal
Clone: [5C1]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3,k
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR