Anti-TLN1 Mouse Monoclonal Antibody

Supplier: Prosci

51-520
10073-268EA 624.31 USD InStock
10073-268
Anti-TLN1 Mouse Monoclonal Antibody
Antibodies
Talin antibody can be used in Western Blot, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

Store Talin antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: TLN1
Clonality: Monoclonal
Clone: [1A11]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3,k
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR