Anti-RTKN Mouse Monoclonal Antibody [clone: [2E5]]

Supplier: Prosci

51-507
10077-002EA 647.66 USD
10077-002
Anti-RTKN Mouse Monoclonal Antibody [clone: [2E5]]
Antibodies
RTKN antibody can be used in Western Blot, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

Store RTKN antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: RTKN
Clonality: Monoclonal
Clone: [2E5]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR