Anti-RPS7 Mouse Monoclonal Antibody

Supplier: Prosci

51-504
10076-996EA 624.31 USD InStock
10076-996
Anti-RPS7 Mouse Monoclonal Antibody
Antibodies
RPS7 antibody can be used in Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

Store RPS7 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: RPS7
Clonality: Monoclonal
Clone: [3G4]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR