Anti-IRF4 Mouse Monoclonal Antibody [clone: [2F2]]

Supplier: Prosci

51-476
10074-766EA 624.31 USD
10074-766
Anti-IRF4 Mouse Monoclonal Antibody [clone: [2F2]]
Antibodies
IRF4 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 ?g/mL, immunofluorescence, and immunoprecipitation.

PBS, pH 7.2

Store IRF4 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: IRF4
Clonality: Monoclonal
Clone: [2F2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR