Anti-IRAK1 Mouse Monoclonal Antibody [clone: [3F7]]

Supplier: ProSci
51-474
10074-762EA 749.24 USD
10074-762
Anti-IRAK1 Mouse Monoclonal Antibody [clone: [3F7]]
Antibodies
IRAK antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 20 ?g/mL.

PBS, pH 7.2

Store IRAK antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: IRAK1
Clonality: Monoclonal
Clone: [3F7]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR