Anti-IDO1 Mouse Monoclonal Antibody [clone: [4D2]]

Supplier: Prosci

51-473
10074-760EA 647.47 USD
10074-760
Anti-IDO1 Mouse Monoclonal Antibody [clone: [4D2]]
Antibodies
IDO antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 20 ?g/mL.

PBS, pH 7.2

Store IDO antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: IDO1
Clonality: Monoclonal
Clone: [4D2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR