Anti-HMGB2 Mouse Monoclonal Antibody [clone: [3D2]]

Supplier: ProSci
51-469
10074-752EA 778.14 USD
10074-752
Anti-HMGB2 Mouse Monoclonal Antibody [clone: [3D2]]
Antibodies
HMGB2 antibody can be used in Western Blot starting at 1 - 2 ?g/mL, and immunohistochemistry starting at 2.5 ?g/mL.

PBS, pH 7.2

Store HMGB2 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: HMGB2
Clonality: Monoclonal
Clone: [3D2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR