Anti-HMGB2 Mouse Monoclonal Antibody [clone: [3C7]]

Supplier: Prosci

51-468
10074-750EA 647.66 USD
10074-750
Anti-HMGB2 Mouse Monoclonal Antibody [clone: [3C7]]
Antibodies
HMGB2 antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

Store HMGB2 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: HMGB2
Clonality: Monoclonal
Clone: [3C7]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR