Anti-HCLS1 Mouse Monoclonal Antibody [clone: [2A8]]

Supplier: Prosci

51-464
10076-974EA 647.66 USD
10076-974
Anti-HCLS1 Mouse Monoclonal Antibody [clone: [2A8]]
Antibodies
HCLS1 antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 5 - 10 ?g/mL.

PBS, pH 7.2

Store HCLS1 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: HCLS1
Clonality: Monoclonal
Clone: [2A8]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR