Anti-HBB Mouse Monoclonal Antibody [clone: [7B12]]

Supplier: ProSci
51-463
10076-972EA 778.14 USD
10076-972
Anti-HBB Mouse Monoclonal Antibody [clone: [7B12]]
Antibodies
HBB antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.2

Store HBB antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: HBB
Clonality: Monoclonal
Clone: [7B12]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR