Anti-GYG1 Mouse Monoclonal Antibody [clone: [3B5]]

Supplier: ProSci
51-462
10076-970EA 749.47 USD
10076-970
Anti-GYG1 Mouse Monoclonal Antibody [clone: [3B5]]
Antibodies
Glycogenin 1 antibody can be used in Western Blot, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

Store Glycogenin 1 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: GYG1
Clonality: Monoclonal
Clone: [3B5]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR