Anti-SFN Mouse Monoclonal Antibody [clone: [3C3]]

Supplier: Prosci

51-459
10076-964EA 647.66 USD
10076-964
Anti-SFN Mouse Monoclonal Antibody [clone: [3C3]]
Antibodies
14-3-3 Sigma antibody can be used in ELISA, Western Blot starting at 1:500 - 1:2000, immunohistochemistry starting at 1:200, and immunofluorescence starting at 1:200 - 1:1000.

PBS, pH 7.2

Store 14-3-3 Sigma antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: SFN
Clonality: Monoclonal
Clone: [3C3]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR