Anti-GART Mouse Monoclonal Antibody [clone: [4D6-1D5]]

Supplier: ProSci
51-456
10076-958EA 749.47 USD
10076-958
Anti-GART Mouse Monoclonal Antibody [clone: [4D6-1D5]]
Antibodies
GART antibody can be used in Western Blot starting at 1 - 2 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

Store GART antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: GART
Clonality: Monoclonal
Clone: [4D6-1D5]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR