Anti-ENO1 Mouse Monoclonal Antibody [clone: [8G8]]

Supplier: ProSci
51-455
10076-956EA 749.47 USD
10076-956
Anti-ENO1 Mouse Monoclonal Antibody [clone: [8G8]]
Antibodies
ENO1 antibody can be used in immunohistochemistry starting at 1/10 - 1/40, and immunohistochemistry in frozen tissues starting at 1/10 - 1/40.

PBS, pH 7.2

Store ENO1 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: ENO1
Clonality: Monoclonal
Clone: [8G8]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR