Anti-DUT Mouse Monoclonal Antibody [clone: [1C9]]

Supplier: ProSci
51-452
10076-950EA 749.47 USD
10076-950
Anti-DUT Mouse Monoclonal Antibody [clone: [1C9]]
Antibodies
DUT antibody can be used in immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.2

Store DUT antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: DUT
Clonality: Monoclonal
Clone: [1C9]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR