Anti-CSE1L Mouse Monoclonal Antibody [clone: [3F8]]

Supplier: Prosci

51-444
10074-742EA 647.66 USD
10074-742
Anti-CSE1L Mouse Monoclonal Antibody [clone: [3F8]]
Antibodies
CSE1L antibody can be used in Western Blot, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.2

Store CSE1L antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: CSE1L
Clonality: Monoclonal
Clone: [3F8]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR