Anti-CDH18 Mouse Monoclonal Antibody [clone: [6F7]]

Supplier: Prosci

51-438
10074-730EA 647.66 USD
10074-730
Anti-CDH18 Mouse Monoclonal Antibody [clone: [6F7]]
Antibodies
Ey-Cadherin antibody can be used in ELISA starting at 1:2000 - 1:10000, Western Blot starting at 1:500 - 1:2000, immunohistochemistry starting at 2.5 ?g/mL, and immunofluorescence.

PBS, pH 7.2

Store Ey-Cadherin antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: CDH18
Clonality: Monoclonal
Clone: [6F7]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR