Anti-CDH1 Mouse Monoclonal Antibody [clone: [3F4]]

Supplier: ProSci
51-437
10074-728EA 836.5 USD
10074-728
Anti-CDH1 Mouse Monoclonal Antibody [clone: [3F4]]
Antibodies
  • E Cadherin antibody can be used in Western Blot starting at 1 - 2 μg/mL, and immunohistochemistry starting at 1 - 2 μg/mL.

Protein A Column

PBS, pH 7.2

Store E Cadherin antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: CDH1
Clonality: Monoclonal
Clone: [3F4]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR