Anti-BUB1B Mouse Monoclonal Antibody [clone: [2G5]]

Supplier: Prosci

51-430
10076-938EA 624.31 USD InStock
10076-938
Anti-BUB1B Mouse Monoclonal Antibody [clone: [2G5]]
Antibodies
  • BubR1 antibody can be used in Western Blot, immunohistochemistry starting at 10 μg/mL, and immunoprecipitation.

Protein A Column

PBS, pH 7.2

Store BubR1 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: BUB1B
Clonality: Monoclonal
Clone: [2G5]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR