Anti-ANXA4 Mouse Monoclonal Antibody [clone: [1D3]]

Supplier: ProSci
51-421
10076-920EA 760.5 USD
10076-920
Anti-ANXA4 Mouse Monoclonal Antibody [clone: [1D3]]
Antibodies
Annexin 4 antibody can be used in ELISA starting at 1:25000, Western Blot starting at 1:500 - 1:2000, and immunohistochemistry starting at 20 - 40 ?g/mL.

PBS, pH 7.2

Store Annexin 4 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: ANXA4
Clonality: Monoclonal
Clone: [1D3]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR