Anti-ALOX15 Mouse Monoclonal Antibody [clone: [3D8]]

Supplier: ProSci
51-420
10076-918EA 760.5 USD
10076-918
Anti-ALOX15 Mouse Monoclonal Antibody [clone: [3D8]]
Antibodies
ALOX15 antibody can be used in Western Blot, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

Store ALOX15 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: ALOX15
Clonality: Monoclonal
Clone: [3D8]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR