Anti-MUC1 Mouse Monoclonal Antibody

Supplier: Prosci

51-418
10076-916EA 593.09 USD
10076-916
Anti-MUC1 Mouse Monoclonal Antibody
Antibodies
MUC1 antibody can be used in ELISA starting at 1:2000 - 1:10000, Western Blot starting at 1:500 - 1:3000, immunohistochemistry starting at 2.5 ?g/mL, and immunofluorescence.

PBS, pH 7.4, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR