Anti-TIMP2 Mouse Monoclonal Antibody [clone: [3A4]]

Supplier: ProSci
51-417
10076-914EA 674.53 USD
10076-914
Anti-TIMP2 Mouse Monoclonal Antibody [clone: [3A4]]
Antibodies
TIMP2 antibody can be used in Western Blot, and immunohistochemistry starting at 2 - 4 ?g/mL.

PBS, 0.08% sodium azide.

Store TIMP2 antibody at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: TIMP2
Clonality: Monoclonal
Clone: [3A4]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Chicken, Zebrafish, Pig, Rabbit, Mouse, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR