Anti-ARNTL Mouse Monoclonal Antibody [clone: [1C5]]

Supplier: Prosci

51-406
10076-892EA 582.9 USD
10076-892
Anti-ARNTL Mouse Monoclonal Antibody [clone: [1C5]]
Antibodies
BMAL1 antibody can be used in immunohistochemistry starting at 2.5 ?g/mL.

0.03% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ARNTL
Clonality: Monoclonal
Clone: [1C5]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR