Anti-A1BG Mouse Monoclonal Antibody [clone: [4B5]]

Supplier: ProSci
51-401
10076-882EA 674.53 USD
10076-882
Anti-A1BG Mouse Monoclonal Antibody [clone: [4B5]]
Antibodies
A1BG antibody can be used in ELISA, Western Blot starting at 0.5 - 2 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

0.03% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: A1BG
Clonality: Monoclonal
Clone: [4B5]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR