Anti-CD69 Mouse Monoclonal Antibody [clone: [8B6]]

Supplier: Prosci

51-399
10076-878EA 561.89 USD
10076-878
Anti-CD69 Mouse Monoclonal Antibody [clone: [8B6]]
Antibodies
CD69 antibody can be used in immunohistochemistry starting at 5 ?g/mL.

0.03% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: [8B6]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR