Anti-BDH1 Mouse Monoclonal Antibody [clone: [1A5]]

Supplier: Prosci

51-391
10076-862EA 582.9 USD
10076-862
Anti-BDH1 Mouse Monoclonal Antibody [clone: [1A5]]
Antibodies
BDH1 antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 5 ?g/mL.

0.03% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: BDH1
Clonality: Monoclonal
Clone: [1A5]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR