Anti-PEG10 Mouse Monoclonal Antibody [clone: [4C10A7]]

Supplier: Prosci

51-373
10076-828EA 582.9 USD
10076-828
Anti-PEG10 Mouse Monoclonal Antibody [clone: [4C10A7]]
Antibodies
PEG10 antibody can be used in ELISA starting at 1:1000, and immunohistochemistry starting at 10 ?g/mL.

0.03% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PEG10
Clonality: Monoclonal
Clone: [4C10A7]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR