Anti-LPA Mouse Monoclonal Antibody [clone: [8F6A9,8H5C5]]

Supplier: ProSci
51-371
10076-824EA 674.53 USD
10076-824
Anti-LPA Mouse Monoclonal Antibody [clone: [8F6A9,8H5C5]]
Antibodies
LPA antibody can be used in Western Blot, and immunohistochemistry starting at 10 ?g/mL.

0.03% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: LPA
Clonality: Monoclonal
Clone: [8F6A9,8H5C5]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR