Anti-BHMT Mouse Monoclonal Antibody [clone: [8C11H5]]

Supplier: ProSci
51-368
10076-818EA 674.53 USD
10076-818
Anti-BHMT Mouse Monoclonal Antibody [clone: [8C11H5]]
Antibodies
BHMT antibody can be used in Western Blot starting at 1:200, immunohistochemistry starting at 10 ?g/mL, and immunoprecipitation.

0.03% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: BHMT
Clonality: Monoclonal
Clone: [8C11H5]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR