Anti-FCER1A Mouse Monoclonal Antibody [clone: [AER-37]

Supplier: Prosci

51-350
10074-712EA 582.9 USD
10074-712
Anti-FCER1A Mouse Monoclonal Antibody [clone: [AER-37]
Antibodies
FCER1A antibody can be used in ELISA, Western Blot starting at 1 ?g/mL, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.2, 150 mM NaCl, 0.09% sodium azide

Store FCER1A antibody at 4 °C. As with all antibodies avoid freeze/thaw cycles

Type: Primary
Antigen: FCER1A
Clonality: Monoclonal
Clone: [AER-37
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR